missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobutyric Acid 99.0+%, TCI America™

Catalog No. p-7116805
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A028225G 25 g
A0282100G 100 g
A0282500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A028225G Supplier TCI America Supplier No. A028225G
Only null left

Chemical Identifiers

CAS 56-12-2
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00008226
InChI Key BTCSSZJGUNDROE-UHFFFAOYSA-N
Synonym 4-aminobutyric acid, gaba, gamma-aminobutyric acid, piperidic acid, piperidinic acid, aminalon, gaballon, gammalon, mielogen, butanoic acid, 4-amino
PubChem CID 119
ChEBI CHEBI:16865
IUPAC Name 4-aminobutanoic acid
SMILES C(CC(=O)O)CN

Specifications

Melting Point 195°C
Color White
Quantity 25 g
Formula Weight 103.12
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminobutyric Acid
EINECSNumber (9)-0114
RTECSNumber ES6300000
TSCA Yes