missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Aminobenzylamine 98.0+%, TCI America™
$325.36 - $1755.75
Chemical Identifiers
| CAS | 4403-71-8 |
|---|---|
| Molecular Formula | C7H10N2 |
| Molecular Weight (g/mol) | 122.171 |
| MDL Number | MFCD00075513 |
| InChI Key | BFWYZZPDZZGSLJ-UHFFFAOYSA-N |
| Synonym | 4-aminobenzylamine, 4-aminomethyl aniline, p-aminobenzylamine, benzenemethanamine, 4-amino, 4-aminomethyl-phenylamine, 4-aminomethyl phenylamine, 4-aminobenzyl amine dihydrochloride, 4-amonobenzylae, p-aminobenzylamin, 4-amino benzylamine |
| PubChem CID | 427814 |
| IUPAC Name | 4-(aminomethyl)aniline |
| SMILES | C1=CC(=CC=C1CN)N |
Chemical Identifiers
| 4403-71-8 | |
| 122.171 | |
| BFWYZZPDZZGSLJ-UHFFFAOYSA-N | |
| 427814 | |
| C1=CC(=CC=C1CN)N |
| C7H10N2 | |
| MFCD00075513 | |
| 4-aminobenzylamine, 4-aminomethyl aniline, p-aminobenzylamine, benzenemethanamine, 4-amino, 4-aminomethyl-phenylamine, 4-aminomethyl phenylamine, 4-aminobenzyl amine dihydrochloride, 4-amonobenzylae, p-aminobenzylamin, 4-amino benzylamine | |
| 4-(aminomethyl)aniline |
Specifications
| 4403-71-8 | |
| C7H10N2 | |
| 25 g | |
| 4-aminobenzylamine, 4-aminomethyl aniline, p-aminobenzylamine, benzenemethanamine, 4-amino, 4-aminomethyl-phenylamine, 4-aminomethyl phenylamine, 4-aminobenzyl amine dihydrochloride, 4-amonobenzylae, p-aminobenzylamin, 4-amino benzylamine | |
| C1=CC(=CC=C1CN)N | |
| 122.171 | |
| 122.17 | |
| Crystalline Lumps |
| White-Yellow | |
| MFCD00075513 | |
| 3259 | |
| BFWYZZPDZZGSLJ-UHFFFAOYSA-N | |
| 4-(aminomethyl)aniline | |
| 427814 | |
| ≥98.0% (GC,T) | |
| 4-Aminobenzylamine |
Safety and Handling
EINECSNumber : (3)-3897
TSCA : No