missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzylamine, 98%

CAS: 4403-71-8 | C7H10N2 | 122.17 g/mol

$100.71 - $100.71

Chemical Identifiers

CAS 4403-71-8
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00075513
InChI Key BFWYZZPDZZGSLJ-UHFFFAOYSA-N
Synonym 4-aminobenzylamine, 4-aminomethyl aniline, p-aminobenzylamine, benzenemethanamine, 4-amino, 4-aminomethyl-phenylamine, 4-aminomethyl phenylamine, 4-aminobenzyl amine dihydrochloride, 4-amonobenzylae, p-aminobenzylamin, 4-amino benzylamine
PubChem CID 427814
IUPAC Name 4-(aminomethyl)aniline
SMILES C1=CC(=CC=C1CN)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC391450050
SDS View Documents
Thermo Scientific Chemicals
391450050
5 g Glass bottle
Each for $100.71
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4403-71-8
122.17
BFWYZZPDZZGSLJ-UHFFFAOYSA-N
427814
C1=CC(=CC=C1CN)N
C7H10N2
MFCD00075513
4-aminobenzylamine, 4-aminomethyl aniline, p-aminobenzylamine, benzenemethanamine, 4-amino, 4-aminomethyl-phenylamine, 4-aminomethyl phenylamine, 4-aminobenzyl amine dihydrochloride, 4-amonobenzylae, p-aminobenzylamin, 4-amino benzylamine
4-(aminomethyl)aniline

Specifications

4403-71-8
100.0
1.0780g/mL
Authentic
Glass bottle
1.6080 to 1.6100
MFCD00075513
1.078
BFWYZZPDZZGSLJ-UHFFFAOYSA-N
4-(aminomethyl)aniline
427814
98%
4-Aminobenzylamine
97.5
Brown
113°C
97.5% min. (GC)
C7H10N2
NH2CH2C6H4NH2
5 g
4-aminobenzylamine, 4-aminomethyl aniline, p-aminobenzylamine, benzenemethanamine, 4-amino, 4-aminomethyl-phenylamine, 4-aminomethyl phenylamine, 4-aminobenzyl amine dihydrochloride, 4-amonobenzylae, p-aminobenzylamin, 4-amino benzylamine
C1=CC(=CC=C1CN)N
122.17
122.17
Oily Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only