missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzylamine, 98%

Catalog No. AC391451000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC391451000 100 g Glass bottle
AC391450050 5 g Glass bottle
AC391450250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC391451000 Supplier Thermo Scientific Chemicals Supplier No. 391451000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4403-71-8 | C7H10N2 | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4403-71-8
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00075513
InChI Key BFWYZZPDZZGSLJ-UHFFFAOYSA-N
Synonym 4-aminobenzylamine, 4-aminomethyl aniline, p-aminobenzylamine, benzenemethanamine, 4-amino, 4-aminomethyl-phenylamine, 4-aminomethyl phenylamine, 4-aminobenzyl amine dihydrochloride, 4-amonobenzylae, p-aminobenzylamin, 4-amino benzylamine
PubChem CID 427814
IUPAC Name 4-(aminomethyl)aniline
SMILES C1=CC(=CC=C1CN)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Brown
Density 1.0780g/mL
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6080 to 1.6100
Linear Formula NH2CH2C6H4NH2
Quantity 100 g
Specific Gravity 1.078
Formula Weight 122.17
Percent Purity 98%
Physical Form Oily Liquid
Chemical Name or Material 4-Aminobenzylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only