missing translation for 'onlineSavingsMsg'
Learn More

(4-Aminobenzyl)phosphonic Acid 98.0+%, TCI America™

Catalog No. p-7118209
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A3072200MG 200 mg
A30721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A3072200MG Supplier TCI America Supplier No. A3072200MG

Chemical Identifiers

CAS 5424-27-1
Molecular Formula C7H10NO3P
Molecular Weight (g/mol) 187.135
MDL Number MFCD00070204
InChI Key NEKHKXMBGWNTOO-UHFFFAOYSA-N
PubChem CID 223978
IUPAC Name (4-aminophenyl)methylphosphonic acid
SMILES C1=CC(=CC=C1CP(=O)(O)O)N

Specifications

Melting Point 325°C
Color White-Yellow
Quantity 200 mg
Formula Weight 187.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (4-Aminobenzyl)phosphonic Acid
TSCA No
Recommended Storage Freezer