missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzyl Cyanide 98.0+%, TCI America™

Catalog No. p-7117547
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16125G 5 g
A161225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A16125G Supplier TCI America Supplier No. A16125G
Only null left

Chemical Identifiers

CAS 3544-25-0
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.17
MDL Number MFCD00007912
InChI Key YCWRFIYBUQBHJI-UHFFFAOYSA-N
Synonym 4-aminobenzyl cyanide, 4-aminophenylacetonitrile, 2-4-aminophenyl acetonitrile, 4-aminophenyl acetonitrile, p-aminobenzyl cyanide, benzeneacetonitrile, 4-amino, 4-aminobenzylcyanide, 4-amino-phenyl-acetonitrile, 2-4-aminophenyl ethanenitrile, 4-aminobenzyl
PubChem CID 77000
IUPAC Name 2-(4-aminophenyl)acetonitrile
SMILES NC1=CC=C(CC#N)C=C1

Specifications

Melting Point 46°C
Quantity 5 g
UN Number 3439
Formula Weight 132.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminobenzyl Cyanide