missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzotrifluoride 98.0+%, TCI America™

Catalog No. p-7116993
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
25 mL
Catalog No. Quantity
A066225ML 25 mL
A0662100ML 100 mL
2 options

Catalog No. A066225ML

Supplier: TCI America A066225ML

Only null left

Chemical Identifiers

CAS 455-14-1
Molecular Formula C7H6F3N
Molecular Weight (g/mol) 161.127
MDL Number MFCD00064396
InChI Key ODGIMMLDVSWADK-UHFFFAOYSA-N
Synonym 4-trifluoromethyl aniline, 4-aminobenzotrifluoride, p-aminobenzotrifluoride, p-trifluoromethylaniline, benzenamine, 4-trifluoromethyl, 4-trifluoromethyl benzenamine, p-trifluoromethyl aniline, aniline, p-trifluoromethyl, alpha,alpha,alpha-trifluoro-p-toluidine, a,a,a-trifluoro-p-toluidine
PubChem CID 9964
ChEBI CHEBI:40750
IUPAC Name 4-(trifluoromethyl)aniline
SMILES C1=CC(=CC=C1C(F)(F)F)N

Specifications

Boiling Point 117°C
Quantity 25 mL
UN Number 2810
Formula Weight 161.13
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Aminobenzotrifluoride
EINECSNumber (3)-0223
RTECSNumber XU9260000
TSCA Yes
Recommended Storage Refrigerator