missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzotrifluoride, 99%

Catalog No. AC208910100
Change view
Click to view available options
Quantity:
10 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC208910100 10 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC208910100 Supplier Thermo Scientific Chemicals Supplier No. 208910100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 455-14-1 | C7H6F3N | 161.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 455-14-1
Molecular Formula C7H6F3N
Molecular Weight (g/mol) 161.13
MDL Number MFCD00064396
InChI Key ODGIMMLDVSWADK-UHFFFAOYSA-N
Synonym 4-trifluoromethyl aniline, 4-aminobenzotrifluoride, p-aminobenzotrifluoride, p-trifluoromethylaniline, benzenamine, 4-trifluoromethyl, 4-trifluoromethyl benzenamine, p-trifluoromethyl aniline, aniline, p-trifluoromethyl, alpha,alpha,alpha-trifluoro-p-toluidine, a,a,a-trifluoro-p-toluidine
PubChem CID 9964
ChEBI CHEBI:40750
IUPAC Name 4-(trifluoromethyl)aniline
SMILES C1=CC(=CC=C1C(F)(F)F)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 5.0°C to 8.0°C
Color Colorless to Yellow
Density 1.2960g/mL
Boiling Point 83.0°C (12.0 mmHg)
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4830 to 1.4850
Linear Formula H2NC6H4CF3
Quantity 10 mL
Beilstein 12, III, 2151
Specific Gravity 1.296
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ether,methanol,ethanol and,aromatics
Formula Weight 161.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Aminobenzotrifluoride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
Causes serious eye damage.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minu
EINECSNumber 207-236-5
RTECSNumber XU9260000
TSCA TSCA

RUO – Research Use Only