missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzophenone, 98%

Catalog No. p-55926
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC103170100 10 g
AC103170250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC103170100 Supplier Thermo Scientific Chemicals Supplier No. 103170100

CAS: 1137-41-3 | C13H11NO | 197.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1137-41-3
Molecular Formula C13H11NO
Molecular Weight (g/mol) 197.24
MDL Number MFCD00007895
InChI Key RBKHNGHPZZZJCI-UHFFFAOYSA-N
Synonym 4-aminobenzophenone, 4-aminophenyl phenyl methanone, 4-benzoylaniline, p-aminobenzophenone, methanone, 4-aminophenyl phenyl, benzophenone, 4-amino, p-benzoylaniline, usaf a-233, 4-aminophenyl-phenylmethanone, 4-aminophenyl-phenyl-methanone
PubChem CID 14346
IUPAC Name (4-aminophenyl)-phenylmethanone
SMILES NC1=CC=C(C=C1)C(=O)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 120°C to 124°C
Color Yellow
Boiling Point 246°C (17.0 hPa)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H4COC6H5
Quantity 10 g
Beilstein 14, 81
Solubility Information Solubility in water: hardly soluble in cold water. Other solubilities: freely soluble in alcohol and ether
Formula Weight 197.24
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Aminobenzophenone, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 214-506-6
RTECSNumber DJ0175000
TSCA TSCA

RUO – Research Use Only