missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzoic acid, 99%

Catalog No. AAA1267318
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1267318 50 g
AAA1267330 250 g
AAA126730B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1267318 Supplier Thermo Scientific Chemicals Supplier No. A1267318
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 150-13-0 | C7H7NO2 | 137.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-Aminobenzoic acid acts as a gastric acid secretion inhibitor. It is employed as an intermediate in the synthesis of folate (Vitamin BX), azo dyes and cross linking agents. It is also useful in biomedical sector for the preparation of local anesthetic and ointments. It is widely used in industries to make pharmaceutical products, flavors and preservatives. It is actively involved as a UV-blocking ingredient in sun tan cosmetics.

Solubility
Slightly soluble in water. Soluble in methanol and diethyl ether.

Notes
Air and light sensitive. Incompatible with ferric salts and oxidizing agents. May discolor on exposure to light and air. Keep the container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 150-13-0
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00007894
InChI Key ALYNCZNDIQEVRV-UHFFFAOYSA-N
Synonym p-aminobenzoic acid, paba, para-aminobenzoic acid, vitamin bx, sunbrella, p-carboxyaniline, 4-carboxyaniline, hachemina, paraminol, benzoic acid, 4-amino
PubChem CID 978
ChEBI CHEBI:30753
IUPAC Name 4-aminobenzoic acid
SMILES NC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point ∼187°C (decomposition)
Density 1.374
Quantity 50 g
Beilstein 471605
Sensitivity Air and light sensitive
Merck Index 14,423
Solubility Information Slightly soluble in water. Soluble in methanol and diethyl ether.
Formula Weight 137.14
Percent Purity 99%
Chemical Name or Material 4-Aminobenzoic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 205-753-0
RTECSNumber DG1400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only