missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzo-2,1,3-thiadiazole, 98%

Catalog No. p-7045171
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0967803 1 g
AAL0967806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0967803 Supplier Thermo Scientific Chemicals Supplier No. L0967803
Only null left

CAS: 767-64-6 | C6H5N3S | 151.187 g/mol

4-Aminobenzo-2,1,3-thiadiazole is used in a study to synthesize and evaluate the inhibitory activity of a series of substituted benzimidazoles and small benzothiadiazoles on rat liver methionine synthase. It was used as potential levelers for Cu plating in submicrometer trenches.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminobenzo-2,1,3-thiadiazole is used in a study to synthesize and evaluate the inhibitory activity of a series of substituted benzimidazoles and small benzothiadiazoles on rat liver methionine synthase. It was used as potential levelers for Cu plating in submicrometer trenches.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.

Chemical Identifiers

CAS 767-64-6
Molecular Formula C6H5N3S
Molecular Weight (g/mol) 151.187
MDL Number MFCD00005810
InChI Key DRLGIZIAMHIQHL-UHFFFAOYSA-N
Synonym 4-aminobenzo-2,1,3-thiadiazole, 4-amino-2,1,3-benzothiadiazole, 4-aminopiazthiole, benzo c 1,2,5 thiadiazol-4-amine, 2,1,3-benzothiadiazole, 4-amino, 7-amino-2,1,3-benzothiadiazole, benzo 1,2,5 thiadiazol-4-ylamine, 2,1,3-benzothiadiazole, 7-amino, 2,1,3-benzothiadiazol-4-ylamine, 2,1,3-benzothiadiazol-4 1h-imine
PubChem CID 69845
IUPAC Name 2,1,3-benzothiadiazol-4-amine
SMILES C1=CC2=NSN=C2C(=C1)N

Specifications

Melting Point 67°C to 69°C
Quantity 1 g
Beilstein 3551
Solubility Information Insoluble in water.
Formula Weight 151.19
Percent Purity 98%
Chemical Name or Material 4-Aminobenzo-2,1,3-thiadiazole
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 212-186-2
RTECSNumber DK9950000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only