accessibility menu, dialog, popup

UserName

CAS RN 767-64-6

IUPAC Name: 2,1,3-benzothiadiazol-4-amine
Synonyms: 4-aminobenzo-2,1,3-thiadiazole
Molecular Weight (g/mol): 151.19
Molecular Formula: C6H5N3S
InChi Key: DRLGIZIAMHIQHL-UHFFFAOYSA-N
SMILES: NC1=CC=CC2=NSN=C12
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)