missing translation for 'onlineSavingsMsg'
Learn More

4'-Aminobenzanilide, 97%

Catalog No. AAB2566314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2566314 25 g
AAB2566306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2566314 Supplier Thermo Scientific Chemicals Supplier No. B2566314

CAS: 17625-83-1 | C13H12N2O | 212.252 g/mol

4'-Aminobenzanilide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field. It has anticonvulsant property.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Aminobenzanilide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field. It has anticonvulsant property.

Solubility
Soluble in DMSO and methanol.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 17625-83-1
Molecular Formula C13H12N2O
Molecular Weight (g/mol) 212.252
MDL Number MFCD00035777
InChI Key GTTFJYUWPUKXJH-UHFFFAOYSA-N
Synonym 4'-aminobenzanilide, n-4-aminophenyl benzamide, benzamide, n-4-aminophenyl, 4,4'-diaminobenzoylanilide, n-4-amino-phenyl-benzamide, n-4-aminophenyl-benzamide, 4-aminobenzanilide, 4/'-aminobenzanilide, acmc-209xfr, n-benzoyl 4-aminoaniline
PubChem CID 87196
IUPAC Name N-(4-aminophenyl)benzamide
SMILES C1=CC=C(C=C1)C(=O)NC2=CC=C(C=C2)N

Specifications

Melting Point 127°C to 131°C
Quantity 25 g
Solubility Information Soluble in DMSO and methanol.
Formula Weight 212.25
Percent Purity 97%
Chemical Name or Material 4'-Aminobenzanilide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 241-603-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only