missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzamide 98.0+%, TCI America™

$57.75 - $373.48

Chemical Identifiers

CAS 2835-68-9
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.154
MDL Number MFCD00007999
InChI Key QIKYZXDTTPVVAC-UHFFFAOYSA-N
Synonym p-aminobenzamide, benzamide, 4-amino, 4-amino-benzamide, p-carbamoylaniline, p-aminobenzoic acid amide, benzamide, p-amino, 4-amino benzamide, unii-77722i6pac, ccris 6792, aminobenzamide-4
PubChem CID 76079
IUPAC Name 4-aminobenzamide
SMILES C1=CC(=CC=C1C(=O)N)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A063125G
View Documents
TCI America
A063125G
25 g N/A
Only null left
 
A0631500G
View Documents
TCI America
A0631500G
500 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2835-68-9
136.154
QIKYZXDTTPVVAC-UHFFFAOYSA-N
76079
C1=CC(=CC=C1C(=O)N)N
C7H8N2O
MFCD00007999
p-aminobenzamide, benzamide, 4-amino, 4-amino-benzamide, p-carbamoylaniline, p-aminobenzoic acid amide, benzamide, p-amino, 4-amino benzamide, unii-77722i6pac, ccris 6792, aminobenzamide-4
4-aminobenzamide

Specifications

2835-68-9
White-Yellow
MFCD00007999
p-aminobenzamide, benzamide, 4-amino, 4-amino-benzamide, p-carbamoylaniline, p-aminobenzoic acid amide, benzamide, p-amino, 4-amino benzamide, unii-77722i6pac, ccris 6792, aminobenzamide-4
C1=CC(=CC=C1C(=O)N)N
136.154
136.15
Crystalline Powder
183°C
C7H8N2O
25 g
QIKYZXDTTPVVAC-UHFFFAOYSA-N
4-aminobenzamide
76079
≥98.0% (HPLC,T)
4-Aminobenzamide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1446

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'