missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzamide, 98+%

Catalog No. aaa1259436
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1259436 500 g
AAA1259414 25 g
AAA1259422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1259436 Supplier Thermo Scientific Chemicals Supplier No. A1259436
Only null left

CAS: 2835-68-9 | C7H8N2O | 136.154 g/mol

4-Aminobenzamide was used as a poly(ADP-ribose)polymerase (PADPRP) inhhibitor to study the death of target cells by cytotoxic effector cells using the nuclear enzyme poly(ADP-ribose)polymerase (PADPRP).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminobenzamide was used as a poly(ADP-ribose)polymerase (PADPRP) inhhibitor to study the death of target cells by cytotoxic effector cells using the nuclear enzyme poly(ADP-ribose)polymerase (PADPRP).

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2835-68-9
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.154
MDL Number MFCD00007999
InChI Key QIKYZXDTTPVVAC-UHFFFAOYSA-N
Synonym p-aminobenzamide, benzamide, 4-amino, 4-amino-benzamide, p-carbamoylaniline, p-aminobenzoic acid amide, benzamide, p-amino, 4-amino benzamide, unii-77722i6pac, ccris 6792, aminobenzamide-4
PubChem CID 76079
IUPAC Name 4-aminobenzamide
SMILES C1=CC(=CC=C1C(=O)N)N

Specifications

Melting Point 182°C to 184°C
Quantity 500 g
Beilstein 2079485
Solubility Information Slightly soluble in water.
Formula Weight 136.15
Percent Purity ≥98%
Chemical Name or Material 4-Aminobenzamide
Hazard Category H315-H319-H335-H500
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-612-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only