missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzaldehyde Polymer, TCI America™

Catalog No. A026025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A026025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A026025G Supplier TCI America Supplier No. A026025G

Chemical Identifiers

CAS 28107-09-7
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.14
MDL Number MFCD00038137
InChI Key VATYWCRQDJIRAI-UHFFFAOYSA-N
PubChem CID 11158
IUPAC Name 4-aminobenzaldehyde
SMILES NC1=CC=C(C=O)C=C1

Specifications

Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminobenzaldehyde Polymer
EINECSNumber (3)-1163
TSCA No