missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Aminoantipyrine Hydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Quantity | 5 g |
| Formula Weight | 239.70 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Aminoantipyrine Hydrochloride [for Biochemical Research] |
Safety and Handling
| EINECSNumber | (9)-0062 |
| TSCA | Yes |