accessibility menu, dialog, popup

UserName

CAS RN 22198-72-7

HClCH3NNOH3CH2N

CAS RN 22198-72-7

IUPAC Name: hydrogen 4-amino-1,5-dimethyl-2-phenyl-2,3-dihydro-1H-pyrazol-3-one chloride
Synonyms: hydrogen aminoantipyrine chloride
Molecular Weight (g/mol): 239.7
Molecular Formula: C11H14ClN3O
InChi Key: UZSCVCWALGRUTR-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN1N(C(=O)C(N)=C1C)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results