missing translation for 'onlineSavingsMsg'
Learn More

4'-Aminoacetanilide 98.0+%, TCI America™

Catalog No. p-7116720
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A010625G 25 g
A0106500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A010625G Supplier TCI America Supplier No. A010625G

Chemical Identifiers

CAS 122-80-5
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.181
MDL Number MFCD00007853
InChI Key CHMBIJAOCISYEW-UHFFFAOYSA-N
Synonym 4'-aminoacetanilide, n-4-aminophenyl acetamide, p-aminoacetanilide, 4-aminoacetanilide, 4-acetamidoaniline, p-amino acetanilide, p-acetamidoaniline, acetparamin, acetamide, n-4-aminophenyl, fourrine a
PubChem CID 31230
IUPAC Name N-(4-aminophenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1)N

Specifications

Melting Point 166°C
Quantity 25 g
Formula Weight 150.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Aminoacetanilide
EINECSNumber (3)-0244
RTECSNumber AD8225000
TSCA Yes