missing translation for 'onlineSavingsMsg'
Learn More

4'-Aminoacetanilide, 95%

Catalog No. p-55601
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC400600250 25 g Glass bottle
AC400601000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC400600250 Supplier Thermo Scientific Chemicals Supplier No. 400600250

CAS: 122-80-5 | C8H10N2O | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 122-80-5
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.18
MDL Number MFCD00007853
InChI Key CHMBIJAOCISYEW-UHFFFAOYSA-N
Synonym 4'-aminoacetanilide, n-4-aminophenyl acetamide, p-aminoacetanilide, 4-aminoacetanilide, 4-acetamidoaniline, p-amino acetanilide, p-acetamidoaniline, acetparamin, acetamide, n-4-aminophenyl, fourrine a
PubChem CID 31230
IUPAC Name N-(4-aminophenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1)N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 159.0°C to 165.0°C
Color Brown to Pink
Boiling Point 267.0°C
Flash Point >150°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula NHCOCH3C6H4NH2
Quantity 25 g
Merck Index 15, 405
Solubility Information Solubility in water: soluble in cold water. Other solubilities: freely soluble in hot water,freely soluble in alcohol,ether
Formula Weight 150.18
Percent Purity 95%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4'-Aminoacetanilide, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement GHS P Statement
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
If eye irritation persi
EINECSNumber 204-576-6
RTECSNumber AD8225000
TSCA TSCA

RUO – Research Use Only