missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-N,N-dimethylbenzenesulfonamide, 97%

Catalog No. p-7032330
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3374703 1 g
AAH3374706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3374703 Supplier Thermo Scientific Chemicals Supplier No. H3374703
Only null left

CAS: 1709-59-7 | C8H12N2O2S | 200.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1709-59-7
Molecular Formula C8H12N2O2S
Molecular Weight (g/mol) 200.26
MDL Number MFCD00031428
InChI Key BABGMPQXLCJMSK-UHFFFAOYSA-N
Synonym n,n-dimethylsulfanilamide, n1-dimethylsulfanilamide, 4-amino-n,n-dimethylbenzenesulphonamide, benzenesulfonamide, 4-amino-n,n-dimethyl, n',n'-dimethylsulfonamide, n 1-dimethylsulfanilamide, p-dimethylsulfamoyl aniline, n1,n1-dimethylsulfanilamide, 4-amino-n,n-dimethyl-benzenesulfonamide, p-amino-n,n-dimethylbenzenesulfonamide
PubChem CID 74369
IUPAC Name 4-amino-N,N-dimethylbenzenesulfonamide
SMILES CN(C)S(=O)(=O)C1=CC=C(N)C=C1

Specifications

Melting Point 168°C to 170°C
Quantity 1 g
Formula Weight 200.26
Percent Purity 97%
Chemical Name or Material 4-Amino-N,N-dimethylbenzenesulfonamide
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 216-970-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only