missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-N,N-dimethylbenzamide, 97+%

Catalog No. p-7033280
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5036403 1 g
AAH5036406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5036403 Supplier Thermo Scientific Chemicals Supplier No. H5036403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6331-71-1 | C9H12N2O | 164.21 g/mol

4-Amino-N,N-dimethylbenzamide is an important raw material and intermediate used in pharmaceuticals, agro based and dye stuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-N,N-dimethylbenzamide is an important raw material and intermediate used in pharmaceuticals, agro based and dye stuff fields.

Solubility
Very soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 6331-71-1
Molecular Formula C9H12N2O
Molecular Weight (g/mol) 164.21
MDL Number MFCD00523648
InChI Key QEPGWLBMAAEBCP-UHFFFAOYSA-N
Synonym benzamide, 4-amino-n,n-dimethyl, 4-amino-n,n-dimethyl-benzamide, n,n-dimethyl 4-aminobenzamide, 4-aminophenyl-n,n-dimethylcarboxamide, 4-dimethylcarbamyl aniline, acmc-1bgt4, 4-dimethylcarbamoyl aniline, n,n-dimethyl-p-aminobenzamide, p-amino-n,n-dimethylbenzamide, n,n-dimethyl-4-aminobenzamide
PubChem CID 235527
IUPAC Name 4-amino-N,N-dimethylbenzamide
SMILES CN(C)C(=O)C1=CC=C(N)C=C1

Specifications

Melting Point 151°C to 153°C
Quantity 1 g
Solubility Information Very soluble in water.
Formula Weight 164.21
Percent Purity ≥97%
Chemical Name or Material 4-Amino-N,N-dimethylbenzamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only