missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-6-hydroxypyrimidine 98.0+%, TCI America™

Catalog No. A24391G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24391G 1 g
A24395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24391G Supplier TCI America Supplier No. A24391G

Chemical Identifiers

CAS 1193-22-2
Molecular Formula C4H5N3O
Molecular Weight (g/mol) 111.10
MDL Number MFCD00051502
InChI Key HFMLLTVIMFEQRE-UHFFFAOYSA-N
Synonym 4-amino-6-hydroxypyrimidine, 6-aminopyrimidin-4-ol, 4-hydroxy-6-aminopyrimidine, 4-pyrimidinol, 6-amino, 4 1h-pyrimidinone, 6-amino, 6-aminopyrimidin-4 3h-one, 4-oxy-6-aminopyrimidine, 6-amino-4-pyrimidinol, 6-amino-3h-pyrimidin-4-one, 6-amino-4 1h-pyrimidinone
PubChem CID 70944
IUPAC Name 6-amino-1,4-dihydropyrimidin-4-one
SMILES NC1=CC(=O)N=CN1

Specifications

Melting Point 265°C
Color White-Yellow
Quantity 1 g
Formula Weight 111.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-6-hydroxypyrimidine