accessibility menu, dialog, popup

UserName

CAS RN 1193-22-2

IUPAC Name: 6-amino-1,4-dihydropyrimidin-4-one
Synonyms: 6-amino-1H-pyrimidin-4-one
Molecular Weight (g/mol): 111.1
Molecular Formula: C4H5N3O
InChi Key: HFMLLTVIMFEQRE-UHFFFAOYSA-N
SMILES: NC1=CC(=O)N=CN1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)