missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-6-hydroxypyrimidine, 98%, Thermo Scientific™
Specifications
Specifications
| Loss on Drying | 1% max. (105°C, 3 hr) |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (HPLC) |
| Quantity | 5g |
| Solubility Information | Solubility in water: slightly soluble. |
| Formula Weight | 111.1 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-Amino-6-hydroxypyrimidine |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and kee |
| EINECSNumber | 214-771-8 |
RUO – Research Use Only