missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-6-hydroxypyrimidine, 98%, Thermo Scientific™

Catalog No. AC459750010
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459750010 1g
AC459750050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC459750010 Supplier Thermo Scientific Chemicals Supplier No. 459750010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1193-22-2
Molecular Formula C4H5N3O
Molecular Weight (g/mol) 111.10
MDL Number MFCD00051502
InChI Key HFMLLTVIMFEQRE-UHFFFAOYSA-N
Synonym 4-amino-6-hydroxypyrimidine, 6-aminopyrimidin-4-ol, 4-hydroxy-6-aminopyrimidine, 4-pyrimidinol, 6-amino, 4 1h-pyrimidinone, 6-amino, 6-aminopyrimidin-4 3h-one, 4-oxy-6-aminopyrimidine, 6-amino-4-pyrimidinol, 6-amino-3h-pyrimidin-4-one, 6-amino-4 1h-pyrimidinone
PubChem CID 70944
IUPAC Name 6-amino-1H-pyrimidin-4-one
SMILES NC1=CC(=O)N=CN1

Specifications

Loss on Drying 1% max. (105°C, 3 hr)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1g
Solubility Information Solubility in water: slightly soluble.
Formula Weight 111.1
Percent Purity 98%
Chemical Name or Material 4-Amino-6-hydroxypyrimidine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 214-771-8

RUO – Research Use Only