missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 1193-22-2 |
|---|---|
| Molecular Formula | C4H5N3O |
| Molecular Weight (g/mol) | 111.10 |
| MDL Number | MFCD00051502 |
| InChI Key | HFMLLTVIMFEQRE-UHFFFAOYSA-N |
| Synonym | 4-amino-6-hydroxypyrimidine, 6-aminopyrimidin-4-ol, 4-hydroxy-6-aminopyrimidine, 4-pyrimidinol, 6-amino, 4 1h-pyrimidinone, 6-amino, 6-aminopyrimidin-4 3h-one, 4-oxy-6-aminopyrimidine, 6-amino-4-pyrimidinol, 6-amino-3h-pyrimidin-4-one, 6-amino-4 1h-pyrimidinone |
| PubChem CID | 70944 |
| IUPAC Name | 6-amino-1H-pyrimidin-4-one |
| SMILES | NC1=CC(=O)N=CN1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC459750010
|
N/A | 1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC459750050
|
N/A | 5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| C4H5N3O | |
| MFCD00051502 | |
| 4-amino-6-hydroxypyrimidine, 6-aminopyrimidin-4-ol, 4-hydroxy-6-aminopyrimidine, 4-pyrimidinol, 6-amino, 4 1h-pyrimidinone, 6-amino, 6-aminopyrimidin-4 3h-one, 4-oxy-6-aminopyrimidine, 6-amino-4-pyrimidinol, 6-amino-3h-pyrimidin-4-one, 6-amino-4 1h-pyrimidinone | |
| 6-amino-1H-pyrimidin-4-one |
Specifications
| 1193-22-2 | |
| Authentic | |
| Glass Bottle | |
| MFCD00051502 | |
| Solubility in water: slightly soluble. | |
| NC1=CC(=O)N=CN1 | |
| 111.10 | |
| 111.1 | |
| 4-Amino-6-hydroxypyrimidine |
| 1% max. (105°C, 3 hr) | |
| 97.5% min. (HPLC) | |
| C4H5N3O | |
| 4-amino-6-hydroxypyrimidine, 6-aminopyrimidin-4-ol, 4-hydroxy-6-aminopyrimidine, 4-pyrimidinol, 6-amino, 4 1h-pyrimidinone, 6-amino, 6-aminopyrimidin-4 3h-one, 4-oxy-6-aminopyrimidine, 6-amino-4-pyrimidinol, 6-amino-3h-pyrimidin-4-one, 6-amino-4 1h-pyrimidinone | |
| HFMLLTVIMFEQRE-UHFFFAOYSA-N | |
| 6-amino-1H-pyrimidin-4-one | |
| 70944 | |
| 98% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
EINECSNumber : 214-771-8
RUO – Research Use Only