missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-6-hydroxy-2-methylpyrimidine Hydrate 98.0+%, TCI America™

Supplier:  TCI America A14705G

 View more versions of this product

Catalog No. A14705G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-Amino-6-hydroxy-2-methylpyrimidine Hydrate
White-Yellow
MFCD00150312
6-Amino-2-methyl-4-pyrimidinol, 6-Amino-2-methyl-3H-pyrimidin-4-one
CC1=NC(=O)C=C(N1)N.O
143.146
125.13
Crystalline Powder
732304-23-3
C5H9N3O2
5 g
DASZPEWSQNRDMO-UHFFFAOYSA-N
6-amino-2-methyl-1H-pyrimidin-4-one;hydrate
45789592
≥98.0% (T)

Chemical Identifiers

732304-23-3
143.146
DASZPEWSQNRDMO-UHFFFAOYSA-N
45789592
CC1=NC(=O)C=C(N1)N.O
C5H9N3O2
MFCD00150312
6-Amino-2-methyl-4-pyrimidinol, 6-Amino-2-methyl-3H-pyrimidin-4-one
6-amino-2-methyl-1H-pyrimidin-4-one;hydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-3814

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions