missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-6-hydroxy-2-methylpyrimidine Hydrate 98.0+%, TCI America™

Catalog No. p-7117458
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14705G 5 g
A147025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14705G Supplier TCI America Supplier No. A14705G
Only null left

Chemical Identifiers

CAS 732304-23-3
Molecular Formula C5H9N3O2
Molecular Weight (g/mol) 143.146
MDL Number MFCD00150312
InChI Key DASZPEWSQNRDMO-UHFFFAOYSA-N
Synonym 6-Amino-2-methyl-4-pyrimidinol, 6-Amino-2-methyl-3H-pyrimidin-4-one
PubChem CID 45789592
IUPAC Name 6-amino-2-methyl-1H-pyrimidin-4-one;hydrate
SMILES CC1=NC(=O)C=C(N1)N.O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 125.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-6-hydroxy-2-methylpyrimidine Hydrate
EINECSNumber (5)-3814
TSCA Yes