missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-6-hydroxy-2-methylpyrimidine dihydrate, 98+%

Catalog No. p-7044629
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0749606 5 g
AAL0749614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0749606 Supplier Thermo Scientific Chemicals Supplier No. L0749606

CAS: 388582-41-0 | C5H11N3O3 | 161.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 388582-41-0
Molecular Formula C5H11N3O3
Molecular Weight (g/mol) 161.16
MDL Number MFCD00150311
InChI Key QJZWHXSXVBZLJI-UHFFFAOYSA-N
Synonym 4-amino-6-hydroxy-2-methylpyrimidine dihydrate, 6-amino-2-methylpyrimidin-4-ol dihydrate, 6-amino-2-methyl-1h-pyrimidin-4-one dihydrate, acmc-1adza, 4 3h-pyrimidinone,6-amino-2-methyl-, hydrate 1:2, 4-amino-6-hydroxy-2-methylpyrimidine dihydrate, 98+%, 6-amino-2-methyl-3-hydropyrimidin-4-one, hydrate, hydrate
PubChem CID 2724727
IUPAC Name 6-amino-2-methyl-1H-pyrimidin-4-one;dihydrate
SMILES O.O.CC1=NC(=O)C=C(N)N1

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 118273
Formula Weight 161.16 (125.13 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material 4-Amino-6-hydroxy-2-methylpyrimidine dihydrate
EINECSNumber 212-182-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only