CAS RN 388582-41-0
IUPAC Name:
6-amino-2-methyl-1,4-dihydropyrimidin-4-one dihydrate
Synonyms:
6-amino-2-methyl-1H-pyrimidin-4-one dihydrate
Molecular Weight (g/mol):
161.16
Molecular Formula:
C5H11N3O3
InChi Key:
QJZWHXSXVBZLJI-UHFFFAOYSA-N
SMILES:
O.O.CC1=NC(=O)C=C(N)N1
1
–
1
of
1
results
1
Promotions Available
4-Amino-6-hydroxy-2-methylpyrimidine dihydrate, 98+%
4-Amino-6-hydroxy-2-methylpyrimidine dihydrate, 98+%
CAS: 388582-41-0 Molecular Formula: C5H11N3O3 Molecular Weight (g/mol): 161.16 MDL Number: MFCD00150311 InChI Key: QJZWHXSXVBZLJI-UHFFFAOYSA-N Synonym: 4-amino-6-hydroxy-2-methylpyrimidine dihydrate,6-amino-2-methylpyrimidin-4-ol dihydrate,6-amino-2-methyl-1h-pyrimidin-4-one dihydrate,acmc-1adza,4 3h-pyrimidinone,6-amino-2-methyl-, hydrate 1:2,4-amino-6-hydroxy-2-methylpyrimidine dihydrate, 98+%,6-amino-2-methyl-3-hydropyrimidin-4-one, hydrate, hydrate PubChem CID: 2724727 IUPAC Name: 6-amino-2-methyl-1H-pyrimidin-4-one;dihydrate SMILES: O.O.CC1=NC(=O)C=C(N)N1