missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-6-hydroxy-2-mercaptopyrimidine Hydrate 98.0+%, TCI America™

Catalog No. A079125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A079125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A079125G Supplier TCI America Supplier No. A079125G
Only null left

Chemical Identifiers

CAS 65802-56-4
Molecular Formula C4H7N3O2S
Molecular Weight (g/mol) 161.179
MDL Number MFCD00052384
InChI Key BWYGMIYEADAIGW-UHFFFAOYSA-N
Synonym 4-amino-6-hydroxy-2-mercaptopyrimidine monohydrate, 6-amino-2-thiouracil monohydrate, 6-amino-2-thioxo-2,3-dihydropyrimidin-4 1h-one hydrate, 2-pyrimidinethiol, 4-amino-6-hydroxy-, monohydrate, 4-amino-6-hydroxy-2-mercaptopyrimidine hydrate, 6-amino-2-sulfanylpyrimidin-4-ol hydrate, 6-amino-2-mercaptopyrimidin-4-ol hydrate, 4 1h-pyrimidinone, 6-amino-2,3-dihydro-2-thioxo-, monohydrate, 6-amino-2-mercapto-4-pyrimidinol monohydrate, 6-amino-2-sulfanylidene-1h-pyrimidin-4-one hydrate
PubChem CID 2723599
IUPAC Name 6-amino-2-sulfanylidene-1H-pyrimidin-4-one;hydrate
SMILES C1=C(NC(=S)NC1=O)N.O

Specifications

Quantity 25 g
Formula Weight 143.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-6-hydroxy-2-mercaptopyrimidine Hydrate
RTECSNumber UW0495000
TSCA Yes