missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-5-fluoro-2-methoxypyrimidine 99.0+%, TCI America™

Catalog No. A21255G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A21255G Supplier TCI America Supplier No. A21255G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1993-63-1
Molecular Formula C5H6FN3O
Molecular Weight (g/mol) 143.12
MDL Number MFCD09264289
InChI Key BYALTIIVASWNSY-UHFFFAOYSA-N
PubChem CID 44630535
IUPAC Name 5-fluoro-2-methoxypyrimidin-4-amine
SMILES COC1=NC=C(F)C(N)=N1

Specifications

Melting Point 193°C
Color White-Yellow
Quantity 5 g
Formula Weight 143.12
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-5-fluoro-2-methoxypyrimidine