accessibility menu, dialog, popup

UserName

H3CONNH2NF

CAS RN 1993-63-1

IUPAC Name: 5-fluoro-2-methoxypyrimidin-4-amine
Synonyms: 5-fluoro-2-methoxypyrimidin-4-amine
Molecular Weight (g/mol): 143.12
Molecular Formula: C5H6FN3O
InChi Key: BYALTIIVASWNSY-UHFFFAOYSA-N
SMILES: COC1=NC=C(F)C(N)=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results