missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-5-chloro-2-ethoxybenzoic Acid 98.0+%, TCI America™

Catalog No. p-7117644
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19415G 5 g
A194125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19415G Supplier TCI America Supplier No. A19415G

Chemical Identifiers

CAS 108282-38-8
Molecular Formula C9H10ClNO3
Molecular Weight (g/mol) 215.63
MDL Number MFCD03840527
InChI Key XWGYOMHQGQZRLC-UHFFFAOYSA-N
PubChem CID 10536658
IUPAC Name 4-amino-5-chloro-2-ethoxybenzoic acid
SMILES CCOC1=CC(N)=C(Cl)C=C1C(O)=O

Specifications

Melting Point 169°C
Color White-Yellow
Quantity 5 g
Formula Weight 215.63
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-5-chloro-2-ethoxybenzoic Acid