missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-4'-methyldiphenyl Ether 98.0+%, TCI America™
Supplier: TCI America A100925G
Specifications
| 4-Amino-4′-methyldiphenyl Ether | |
| 122°C | |
| MFCD00059858 | |
| 4-Aminophenyl p-Tolyl Ether, 4-(p-Tolyloxy)aniline | |
| CC1=CC=C(C=C1)OC2=CC=C(C=C2)N | |
| 199.253 | |
| 199.25 | |
| Crystalline Powder |
| 41295-20-9 | |
| C13H13NO | |
| 25 g | |
| VPCGOYHSWIYEMO-UHFFFAOYSA-N | |
| 4-(4-methylphenoxy)aniline | |
| 94519 | |
| ≥98.0% (T) |
Chemical Identifiers
| 41295-20-9 | |
| 199.253 | |
| VPCGOYHSWIYEMO-UHFFFAOYSA-N | |
| 94519 | |
| CC1=CC=C(C=C1)OC2=CC=C(C=C2)N |
| C13H13NO | |
| MFCD00059858 | |
| 4-Aminophenyl p-Tolyl Ether, 4-(p-Tolyloxy)aniline | |
| 4-(4-methylphenoxy)aniline |
Safety and Handling
TSCA : No