missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-4'-dimethylaminoazobenzene 97.0+%, TCI America™
$216.24 - $745.62
Chemical Identifiers
| CAS | 539-17-3 |
|---|---|
| Molecular Formula | C14H16N4 |
| Molecular Weight (g/mol) | 240.31 |
| MDL Number | MFCD00010204 |
| InChI Key | BVRIUXYMUSKBHG-UHFFFAOYSA-N |
| Synonym | n,n-dimethyl-4,4'-azodianiline, 4'-amino-dab, c.i. disperse black 3, adab, acetile diazo black n, acetile diazo black r, 4-amino-4'-dimethylaminoazobenzene, supracet diazo black a, diazo nero microsetile g, microsetile diazo black g |
| PubChem CID | 10872 |
| IUPAC Name | 4-[2-(4-aminophenyl)diazen-1-yl]-N,N-dimethylaniline |
| SMILES | CN(C)C1=CC=C(C=C1)N=NC1=CC=C(N)C=C1 |
Chemical Identifiers
| 539-17-3 | |
| 240.31 | |
| BVRIUXYMUSKBHG-UHFFFAOYSA-N | |
| 10872 | |
| CN(C)C1=CC=C(C=C1)N=NC1=CC=C(N)C=C1 |
| C14H16N4 | |
| MFCD00010204 | |
| n,n-dimethyl-4,4'-azodianiline, 4'-amino-dab, c.i. disperse black 3, adab, acetile diazo black n, acetile diazo black r, 4-amino-4'-dimethylaminoazobenzene, supracet diazo black a, diazo nero microsetile g, microsetile diazo black g | |
| 4-[2-(4-aminophenyl)diazen-1-yl]-N,N-dimethylaniline |
Specifications
| 539-17-3 | |
| Red-Yellow | |
| MFCD00010204 | |
| n,n-dimethyl-4,4'-azodianiline, 4'-amino-dab, c.i. disperse black 3, adab, acetile diazo black n, acetile diazo black r, 4-amino-4'-dimethylaminoazobenzene, supracet diazo black a, diazo nero microsetile g, microsetile diazo black g | |
| CN(C)C1=CC=C(C=C1)N=NC1=CC=C(N)C=C1 | |
| 240.31 | |
| 240.31 | |
| Crystalline Powder |
| 189°C | |
| C14H16N4 | |
| 1 g | |
| BVRIUXYMUSKBHG-UHFFFAOYSA-N | |
| 4-[2-(4-aminophenyl)diazen-1-yl]-N,N-dimethylaniline | |
| 10872 | |
| ≥97.0% (HPLC,T) | |
| 4-Amino-4′-dimethylaminoazobenzene |
Safety and Handling
EINECSNumber : (5)-4849
RTECSNumber : BX5020000
TSCA : Yes