CAS RN 539-17-3
CAS RN 539-17-3
IUPAC Name:
4-[2-(4-aminophenyl)diazen-1-yl]-N,N-dimethylaniline
Synonyms:
4-[2-(4-aminophenyl)diazen-1-yl]-N,N-dimethylaniline
Molecular Weight (g/mol):
240.31
Molecular Formula:
C14H16N4
InChi Key:
BVRIUXYMUSKBHG-UHFFFAOYSA-N
SMILES:
CN(C)C1=CC=C(C=C1)N=NC1=CC=C(N)C=C1
1
–
1
of
1
results
1
4-Amino-4'-dimethylaminoazobenzene 97.0+%, TCI America™
CAS: 539-17-3 Molecular Formula: C14H16N4 Molecular Weight (g/mol): 240.31 MDL Number: MFCD00010204 InChI Key: BVRIUXYMUSKBHG-UHFFFAOYSA-N Synonym: n,n-dimethyl-4,4'-azodianiline,4'-amino-dab,c.i. disperse black 3,adab,acetile diazo black n,acetile diazo black r,4-amino-4'-dimethylaminoazobenzene,supracet diazo black a,diazo nero microsetile g,microsetile diazo black g PubChem CID: 10872 IUPAC Name: 4-[2-(4-aminophenyl)diazen-1-yl]-N,N-dimethylaniline SMILES: CN(C)C1=CC=C(C=C1)N=NC1=CC=C(N)C=C1