missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-4'-chlorobiphenyl, 97%

Catalog No. AC444460010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC444460010 1 g
AC444460050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC444460010 Supplier Thermo Scientific Chemicals Supplier No. 444460010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 135-68-2 | C12H10ClN | 203.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 135-68-2
Molecular Formula C12H10ClN
Molecular Weight (g/mol) 203.67
InChI Key OREQWMWYRYXCDF-UHFFFAOYSA-N
Synonym 4-amino-4'-chlorobiphenyl, 4'-chloro-1,1'-biphenyl-4-amine, 4'-chlorobiphenyl-4-ylamine, 4'-chlorobiphenyl-4-amine, 4-amino-4'-chlorodiphenyl, 1,1'-biphenyl-4-amine,4'-chloro, unii-c8tzy038qb, 4-4-chlorophenyl aniline, 4-amino-4'-chloro-biphenyl, 4-biphenylamine, 4'-chloro
PubChem CID 8675
IUPAC Name 4-(4-chlorophenyl)aniline
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)Cl)N

Specifications

Melting Point 127°C to 131°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Merck Index 15,43
Formula Weight 203.67
Percent Purity 97%
Chemical Name or Material 4-Amino-4'-chlorobiphenyl
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 205-211-3

RUO – Research Use Only