missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-(trifluoromethyl)benzoic acid, 98%

Catalog No. p-7031417
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3248703 1 g
AAH3248709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3248703 Supplier Thermo Scientific Chemicals Supplier No. H3248703
Only null left

CAS: 400-76-0 | C8H6F3NO2 | 205.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 400-76-0
Molecular Formula C8H6F3NO2
Molecular Weight (g/mol) 205.14
MDL Number MFCD03407960
InChI Key NPPPORJZPNJXNQ-UHFFFAOYSA-N
Synonym 4-amino-3-trifluoromethyl benzoic acid, 4-amino-3-trifluoromethylbenzioc acid, rarechem al bo 0877, 4-amino-5-trifluoromethylbenzoic acid, 4-amino-3-trifluoromethyl-benzoic acid, benzoic acid, 4-amino-3-trifluoromethyl, 4-amino-3-trifluoromethyl benzoic acid 3-trifluoromethyl-4-aminobenzoic acid, 4-amino-3-trifluoromethylbenzoicacid, pubchem2211
PubChem CID 3254286
IUPAC Name 4-amino-3-(trifluoromethyl)benzoic acid
SMILES NC1=C(C=C(C=C1)C(O)=O)C(F)(F)F

Specifications

Melting Point 212°C to 215°C
Quantity 1 g
Beilstein 3291493
Formula Weight 205.14
Percent Purity 98%
Chemical Name or Material 4-Amino-3-(trifluoromethyl)benzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only