missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-nitrobenzoic Acid 95.0+%, TCI America™

Catalog No. p-7117421
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A141725G 25 g
A1417100G 100 g
A1417500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A141725G Supplier TCI America Supplier No. A141725G
Only null left

Chemical Identifiers

CAS 1588-83-6
Molecular Formula C7H6N2O4
Molecular Weight (g/mol) 182.135
MDL Number MFCD00017009
InChI Key ZZNAYFWAXZJITH-UHFFFAOYSA-N
Synonym 4-amino-3-nitrobenzoic acid, 3-nitro-4-aminobenzoic acid, benzoic acid, 4-amino-3-nitro, 4-carboxy-2-nitroaniline, 3-nitro-4-aminobenozic acid, 4-amino-3-nitro-benzoic acid, 4-amino-3-nitrobenzoicacid, pubchem4982, rarechem al bo 0261, 2-nitro-4-carboxyaniline
PubChem CID 74115
SMILES C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])N

Specifications

Color Yellow
Quantity 25 g
Formula Weight 182.14
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-3-nitrobenzoic Acid