missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-nitrobenzoic acid, 98%, Thermo Scientific™

Catalog No. AC400880050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC400880050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC400880050 Supplier Thermo Scientific Chemicals Supplier No. 400880050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1588-83-6
Molecular Formula C7H6N2O4
Molecular Weight (g/mol) 182.14
MDL Number MFCD00017009
InChI Key ZZNAYFWAXZJITH-UHFFFAOYSA-N
Synonym 4-amino-3-nitrobenzoic acid, 3-nitro-4-aminobenzoic acid, benzoic acid, 4-amino-3-nitro, 4-carboxy-2-nitroaniline, 3-nitro-4-aminobenozic acid, 4-amino-3-nitro-benzoic acid, 4-amino-3-nitrobenzoicacid, pubchem4982, rarechem al bo 0261, 2-nitro-4-carboxyaniline
PubChem CID 74115
SMILES C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 290°C
Color Ochre to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: negligible in water
Formula Weight 182.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-3-nitrobenzoic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-453-4

RUO – Research Use Only