missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-methylbenzonitrile 97.0+%, TCI America™

Catalog No. p-7117734
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21071G 1 g
A21075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21071G Supplier TCI America Supplier No. A21071G
Only null left

Chemical Identifiers

CAS 78881-21-7
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.166
MDL Number MFCD02093969
InChI Key MBZDCUMFFPWLTJ-UHFFFAOYSA-N
Synonym 3-methyl-4-aminobenzonitrile, 4-cyano-o-toluidine, 2-amino-5-cyanotoluene, 4-cyano-2-methylaniline, 4-amino-3-methyl-benzonitrile, benzonitrile, 4-amino-3-methyl, pubchem4667, 4-cyano-2-methyianiline, 2-methyl-4-cyanoaniline, acmc-1bc2f
PubChem CID 7010316
IUPAC Name 4-amino-3-methylbenzonitrile
SMILES CC1=C(C=CC(=C1)C#N)N

Specifications

Melting Point 93°C
Quantity 1 g
UN Number 3439
Formula Weight 132.17
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-3-methylbenzonitrile