missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-methylbenzonitrile, 98%

Catalog No. AAL1786803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1786803 1 g
AAL1786806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1786803 Supplier Thermo Scientific Chemicals Supplier No. L1786803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 78881-21-7 | C8H8N2 | 132.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 78881-21-7
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.166
MDL Number MFCD02093969
InChI Key MBZDCUMFFPWLTJ-UHFFFAOYSA-N
Synonym 3-methyl-4-aminobenzonitrile, 4-cyano-o-toluidine, 2-amino-5-cyanotoluene, 4-cyano-2-methylaniline, 4-amino-3-methyl-benzonitrile, benzonitrile, 4-amino-3-methyl, pubchem4667, 4-cyano-2-methyianiline, 2-methyl-4-cyanoaniline, acmc-1bc2f
PubChem CID 7010316
IUPAC Name 4-amino-3-methylbenzonitrile
SMILES CC1=C(C=CC(=C1)C#N)N

Specifications

Melting Point 93°C to 94°C
Quantity 1 g
UN Number UN3439
Beilstein 3235728
Formula Weight 132.17
Percent Purity ≥98%
Chemical Name or Material 4-Amino-3-methylbenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only