missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-3-methoxybenzoic Acid 98.0+%, TCI America™
$186.41 - $741.89
Chemical Identifiers
| CAS | 2486-69-3 |
|---|---|
| Molecular Formula | C8H9NO3 |
| Molecular Weight (g/mol) | 167.164 |
| MDL Number | MFCD00016539 |
| InChI Key | JNFGLYJROFAOQP-UHFFFAOYSA-N |
| Synonym | 4-amino-3-methoxy-benzoic acid, 3-methoxy-4-aminobenzoic acid, 4-amino-m-anisic acid, benzoic acid, 4-amino-3-methoxy, o-anisidine-4-carboxylic acid, 4-amino-3-methoxy benzoic acid, timtec-bb sbb008622, 4-amino-3-methoxybenzoic, 4-amino-3-methoxybenzoic acid, 4-amino-3-methoxybenzoicacid |
| PubChem CID | 288057 |
| IUPAC Name | 4-amino-3-methoxybenzoic acid |
| SMILES | COC1=C(C=CC(=C1)C(=O)O)N |
Chemical Identifiers
| 2486-69-3 | |
| 167.164 | |
| JNFGLYJROFAOQP-UHFFFAOYSA-N | |
| 288057 | |
| COC1=C(C=CC(=C1)C(=O)O)N |
| C8H9NO3 | |
| MFCD00016539 | |
| 4-amino-3-methoxy-benzoic acid, 3-methoxy-4-aminobenzoic acid, 4-amino-m-anisic acid, benzoic acid, 4-amino-3-methoxy, o-anisidine-4-carboxylic acid, 4-amino-3-methoxy benzoic acid, timtec-bb sbb008622, 4-amino-3-methoxybenzoic, 4-amino-3-methoxybenzoic acid, 4-amino-3-methoxybenzoicacid | |
| 4-amino-3-methoxybenzoic acid |
Specifications
| 2486-69-3 | |
| C8H9NO3 | |
| 5 g | |
| JNFGLYJROFAOQP-UHFFFAOYSA-N | |
| 4-amino-3-methoxybenzoic acid | |
| 288057 | |
| ≥98.0% (HPLC,T) | |
| 4-Amino-3-methoxybenzoic Acid |
| 184°C | |
| MFCD00016539 | |
| 4-amino-3-methoxy-benzoic acid, 3-methoxy-4-aminobenzoic acid, 4-amino-m-anisic acid, benzoic acid, 4-amino-3-methoxy, o-anisidine-4-carboxylic acid, 4-amino-3-methoxy benzoic acid, timtec-bb sbb008622, 4-amino-3-methoxybenzoic, 4-amino-3-methoxybenzoic acid, 4-amino-3-methoxybenzoicacid | |
| COC1=C(C=CC(=C1)C(=O)O)N | |
| 167.164 | |
| 167.16 | |
| Crystalline Powder |
Safety and Handling
TSCA : No