missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-methoxybenzoic acid, 95%

Catalog No. AAH2809803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2809803 1 g
AAH2809806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2809803 Supplier Thermo Scientific Chemicals Supplier No. H2809803

CAS: 2486-69-3 | C8H9NO3 | 167.164 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2486-69-3
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00016539
InChI Key JNFGLYJROFAOQP-UHFFFAOYSA-N
Synonym 4-amino-3-methoxy-benzoic acid, 3-methoxy-4-aminobenzoic acid, 4-amino-m-anisic acid, benzoic acid, 4-amino-3-methoxy, o-anisidine-4-carboxylic acid, 4-amino-3-methoxy benzoic acid, timtec-bb sbb008622, 4-amino-3-methoxybenzoic, 4-amino-3-methoxybenzoic acid, 4-amino-3-methoxybenzoicacid
PubChem CID 288057
IUPAC Name 4-amino-3-methoxybenzoic acid
SMILES COC1=C(C=CC(=C1)C(=O)O)N

Specifications

Melting Point 186°C to 188°C
Quantity 1 g
Formula Weight 167.16
Percent Purity 95%
Chemical Name or Material 4-Amino-3-methoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only