missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-3-iodobenzonitrile 98.0+%, TCI America™
$149.13 - $464.14
Chemical Identifiers
| CAS | 33348-34-4 |
|---|---|
| Molecular Formula | C7H5IN2 |
| Molecular Weight (g/mol) | 244.035 |
| MDL Number | MFCD04039964 |
| InChI Key | UOWVTQFTEAYDLM-UHFFFAOYSA-N |
| Synonym | 4-cyano-2-iodoaniline, 1-amino-2-iodo-4-cyanobenzene, benzonitrile, 4-amino-3-iodo, 4-amino-2-iodobenzonitrile 98, 4-amino-3-iodobenzenecarbonitrile, 2-iodo-4-cyanoaniline, 4-cyano-6-iodoaniline, pubchem18486, acmc-20a6lw, 3-iodo-4-aminobenzonitrile |
| PubChem CID | 4416395 |
| IUPAC Name | 4-amino-3-iodobenzonitrile |
| SMILES | C1=CC(=C(C=C1C#N)I)N |
Chemical Identifiers
| 33348-34-4 | |
| 244.035 | |
| UOWVTQFTEAYDLM-UHFFFAOYSA-N | |
| 4416395 | |
| C1=CC(=C(C=C1C#N)I)N |
| C7H5IN2 | |
| MFCD04039964 | |
| 4-cyano-2-iodoaniline, 1-amino-2-iodo-4-cyanobenzene, benzonitrile, 4-amino-3-iodo, 4-amino-2-iodobenzonitrile 98, 4-amino-3-iodobenzenecarbonitrile, 2-iodo-4-cyanoaniline, 4-cyano-6-iodoaniline, pubchem18486, acmc-20a6lw, 3-iodo-4-aminobenzonitrile | |
| 4-amino-3-iodobenzonitrile |
Specifications
| 33348-34-4 | |
| Yellow | |
| MFCD04039964 | |
| 3439 | |
| UOWVTQFTEAYDLM-UHFFFAOYSA-N | |
| 4-amino-3-iodobenzonitrile | |
| 4416395 | |
| ≥98.0% (GC) | |
| 4-Amino-3-iodobenzonitrile |
| 115°C | |
| C7H5IN2 | |
| 1 g | |
| 4-cyano-2-iodoaniline, 1-amino-2-iodo-4-cyanobenzene, benzonitrile, 4-amino-3-iodo, 4-amino-2-iodobenzonitrile 98, 4-amino-3-iodobenzenecarbonitrile, 2-iodo-4-cyanoaniline, 4-cyano-6-iodoaniline, pubchem18486, acmc-20a6lw, 3-iodo-4-aminobenzonitrile | |
| C1=CC(=C(C=C1C#N)I)N | |
| 244.035 | |
| 244.04 | |
| Crystalline Powder |
Safety and Handling
TSCA : No