missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-iodobenzonitrile 98.0+%, TCI America™

Catalog No. A27475G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27475G 5 g
A27471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27475G Supplier TCI America Supplier No. A27475G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 33348-34-4
Molecular Formula C7H5IN2
Molecular Weight (g/mol) 244.035
MDL Number MFCD04039964
InChI Key UOWVTQFTEAYDLM-UHFFFAOYSA-N
Synonym 4-cyano-2-iodoaniline, 1-amino-2-iodo-4-cyanobenzene, benzonitrile, 4-amino-3-iodo, 4-amino-2-iodobenzonitrile 98, 4-amino-3-iodobenzenecarbonitrile, 2-iodo-4-cyanoaniline, 4-cyano-6-iodoaniline, pubchem18486, acmc-20a6lw, 3-iodo-4-aminobenzonitrile
PubChem CID 4416395
IUPAC Name 4-amino-3-iodobenzonitrile
SMILES C1=CC(=C(C=C1C#N)I)N

Specifications

Melting Point 115°C
Color Yellow
Quantity 5 g
UN Number 3439
Formula Weight 244.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-3-iodobenzonitrile