missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-hydroxy-1-naphthalenesulfonic acid, For Spectrophotometric Det. of Si, ≥95.0%, MilliporeSigma™ Supelco™

Catalog No. 111012655
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012655 25 g
111012656 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111012655 Supplier Merck Emd Millipore Supplier No. 0875125G

4-Amino-3-hydroxy-1-naphthalenesulfonic acid is an aromatic amino-sulfonic acid.

Chemical Identifiers

MDL Number MFCD00004019
Synonym 1-Amino-2-naphthol-4-sulfonic acid

Specifications

Melting Point 290°C
CAS 116-63-2
Linear Formula H2 NC10H5(OH)SO3 H
Quantity 25 g
Formula Weight 239.25
Percent Purity ≥95.0% (CHN); ≥95.0%