missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 99%

Catalog No. AAB2433914
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2433914 25 g
AAB2433922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2433914 Supplier Thermo Scientific Chemicals Supplier No. B2433914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 116-63-2 | C10H9NO4S | 239.245 g/mol

4-Amino-3-hydroxy-1-naphthalenesulfonic acid is used as mordant dyes and acid dyes intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-3-hydroxy-1-naphthalenesulfonic acid is used as mordant dyes and acid dyes intermediate.

Solubility
Soluble in bases. Insoluble in water.

Notes
Store away from strong oxidizers, strong bases, acid anhydrides, acid chlorides.

Chemical Identifiers

CAS 116-63-2
Molecular Formula C10H9NO4S
Molecular Weight (g/mol) 239.245
MDL Number MFCD00004019
InChI Key RXCMFQDTWCCLBL-UHFFFAOYSA-N
Synonym 1-amino-2-naphthol-4-sulfonic acid, 4-amino-3-hydroxy-1-naphthalenesulfonic acid, 1-amino-4-sulfo-2-naphthol, 1,2,4-acid, unii-w46sk3p33g, 1-naphthalenesulfonic acid, 4-amino-3-hydroxy, 4-amino-3-hydroxynaphthalene-1-sulphonic acid, 4-amino-3-hydroxynaphthalenesulfonic acid, 1,2,4-aminonaphtholsulfonic acid, 2-hydroxy-4-sulfo-1-naphthylamine
PubChem CID 8316
ChEBI CHEBI:19024
IUPAC Name 4-amino-3-hydroxynaphthalene-1-sulfonic acid
SMILES C1=CC=C2C(=C1)C(=CC(=C2N)O)S(=O)(=O)O

Specifications

Melting Point ∼295°C (decomposition)
Quantity 25 g
Beilstein 2697469
Merck Index 14,454
Solubility Information Soluble in bases. Insoluble in water.
Formula Weight 239.25
Percent Purity 99%
Chemical Name or Material 4-Amino-3-hydroxy-1-naphthalenesulfonic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 204-147-3
RTECSNumber QK1292000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only